5-Piperidin-3-yl-7-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride

Supplier: Matrix Scientific

041093-250MG
101850-786EA 352.31 USD
101850-786
5-Piperidin-3-yl-7-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride
5-Piperidin-3-yl-7-trifluoromethylimidazo[1,2-a]pyrimidine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₁₅Cl₂F₃N₄
MW: 343.18 g/mol
MDL Number: MFCD12028538
CAS Number: 1185299-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR