2-Methoxy-N-(2-piperidin-2-ylethyl)acetamide hydrochloride

Supplier: Matrix Scientific

041090-250MG
101850-780EA 476.92 USD
101850-780
2-Methoxy-N-(2-piperidin-2-ylethyl)acetamide hydrochloride
2-Methoxy-N-(2-piperidin-2-ylethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD12028535
CAS Number: 1185301-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR