2-(4-Methyl-1H-pyrazol-3-yl)piperidine dihydrochloride

Supplier: Matrix Scientific

041087-250MG
101850-774EA 541.54 USD
101850-774
2-(4-Methyl-1H-pyrazol-3-yl)piperidine dihydrochloride
2-(4-Methyl-1H-pyrazol-3-yl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₉H₁₇Cl₂N₃
MDL Number: MFCD12028532
CAS Number: 1185303-93-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR