4-(4-Methyl-4H-[1,2,4]triazol-3-yl)piperidine dihydrochloride

Supplier: Matrix Scientific

041084-250MG
101850-768EA 476.92 USD
101850-768
4-(4-Methyl-4H-[1,2,4]triazol-3-yl)piperidine dihydrochloride
4-(4-Methyl-4H-[1,2,4]triazol-3-yl)piperidine dihydrochloride

Hazard: Irritant

Formula: C₈H₁₆Cl₂N₄
MDL Number: MFCD12028529
CAS Number: 1185301-61-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR