N-(2-Imidazol-1-ylethyl)-2-piperidin-4-ylacetamide hydrochloride

Supplier: Matrix Scientific

041083-250MG
101850-766EA 476.92 USD
101850-766
N-(2-Imidazol-1-ylethyl)-2-piperidin-4-ylacetamide hydrochloride
N-(2-Imidazol-1-ylethyl)-2-piperidin-4-ylacetamide hydrochloride

Hazard: Irritant

Formula: C₁₂H₂₁ClN₄O
MW: 272.78 g/mol
MDL Number: MFCD12028528
CAS Number: 1185171-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR