Morpholine-2-carboxylic acid(1-ethyl-3-methyl-1H-pyrazol-4-yl)amide dihydrochloride

Supplier: Matrix Scientific

041082-250MG
101850-764EA 476.92 USD
101850-764
Morpholine-2-carboxylic acid(1-ethyl-3-methyl-1H-pyrazol-4-yl)amide dihydrochloride
Morpholine-2-carboxylic acid(1-ethyl-3-methyl-1H-pyrazol-4-yl)amide dihydrochloride

Hazard: Irritant

Formula: C₁₁H₂₀Cl₂N₄O₂
MW: 311.21 g/mol
MDL Number: MFCD12028527
CAS Number: 1185126-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR