1-Benzoylpyrrolidine-3-carboxylic acid

Supplier: Matrix Scientific

041080-250MG
101850-760EA 476.92 USD
101850-760
1-Benzoylpyrrolidine-3-carboxylic acid
1-Benzoylpyrrolidine-3-carboxylic acid

Hazard: Irritant

Formula: C₁₂H₁₃NO₃
Melting Pt: 111…113 °C
MDL Number: MFCD12028525
CAS Number: 113558-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR