Pyrrolidine-3-carboxylic acid (2-piperidin-1-ylethyl)amide

Supplier: Matrix Scientific

041075-250MG
101850-750EA 476.92 USD
101850-750
Pyrrolidine-3-carboxylic acid (2-piperidin-1-ylethyl)amide
Pyrrolidine-3-carboxylic acid (2-piperidin-1-ylethyl)amide

Hazard: Irritant

Formula: C₁₂H₂₃N₃O
MW: 225.34 g/mol
MDL Number: MFCD12028521
CAS Number: 1171917-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR