2-Methyl-6,7,8,9-tetrahydro-5H-1,4,7,10a-tetraazacyclohepta[f]inden-10-ol hydrochloride

Supplier: Matrix Scientific

041073-250MG
101850-746EA 476.92 USD
101850-746
2-Methyl-6,7,8,9-tetrahydro-5H-1,4,7,10a-tetraazacyclohepta[f]inden-10-ol hydrochloride
2-Methyl-6,7,8,9-tetrahydro-5H-1,4,7,10a-tetraazacyclohepta[f]inden-10-ol hydrochloride

Hazard: Irritant

Formula: C₁₁H₁₅ClN₄O
MDL Number: MFCD12028519
CAS Number: 1185295-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR