2-Piperidin-4-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride
Supplier: Matrix Scientific
041071-250MG
101850-742EA
476.92
USD
101850-742
2-Piperidin-4-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride
2-Piperidin-4-yl-1H-pyrrolo[2,3-b]pyridine dihydrochloride
Hazard: Irritant
Formula:
C₁₂H₁₇Cl₂N₃ |
MDL Number:
MFCD12028517 CAS Number: 1185295-25-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...