N-(2-Piperidin-2-ylethyl)benzamide hydrochloride

Supplier: Matrix Scientific

041065-500MG
101850-730EA 526.15 USD
101850-730
N-(2-Piperidin-2-ylethyl)benzamide hydrochloride
N-(2-Piperidin-2-ylethyl)benzamide hydrochloride

Hazard: Irritant

Formula: C₁₄H₂₁ClN₂O
MW: 268.79 g/mol
MDL Number: MFCD12028512
CAS Number: 1185303-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR