2-(1-Benzylpiperidin-2-yl)ethylamine dihydrobromide

Supplier: Matrix Scientific

041064-500MG
101850-728EA 526.15 USD
101850-728
2-(1-Benzylpiperidin-2-yl)ethylamine dihydrobromide
2-(1-Benzylpiperidin-2-yl)ethylamine dihydrobromide

Hazard: Irritant

Formula: C₁₄H₂₄Br₂N₂
MW: 380.18 g/mol
MDL Number: MFCD12028511
CAS Number: 1185079-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR