2-[1,3,4]Oxadiazol-2-ylpiperidine hydrochloride

Supplier: Matrix Scientific

041063-500MG
101850-726EA 526.15 USD
101850-726
2-[1,3,4]Oxadiazol-2-ylpiperidine hydrochloride
2-[1,3,4]Oxadiazol-2-ylpiperidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₂ClN₃O
MDL Number: MFCD12028510
CAS Number: 1185011-85-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR