1-[(5-Methylisoxazol-3-ylcarbamoyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Supplier: Matrix Scientific
041059-500MG
101850-718EA
526.15
USD
101850-718
1-[(5-Methylisoxazol-3-ylcarbamoyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid
1-[(5-Methylisoxazol-3-ylcarbamoyl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₂H₁₁N₃O₅ |
MDL Number:
MFCD12028506 CAS Number: 1171917-11-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...