[5-(4,5-Dimethylthiazol-2-ylcarbamoyl)-2-oxo-2H-pyridin-1-yl]acetic acid

Supplier: Matrix Scientific

041055-500MG
101850-710EA 407.69 USD
101850-710
[5-(4,5-Dimethylthiazol-2-ylcarbamoyl)-2-oxo-2H-pyridin-1-yl]acetic acid
[5-(4,5-Dimethylthiazol-2-ylcarbamoyl)-2-oxo-2H-pyridin-1-yl]acetic acid

Hazard: Irritant

Formula: C₁₃H₁₃N₃O₄S
MDL Number: MFCD12028504
CAS Number: 1171917-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR