(5-Cyclohexylcarbamoyl-2-oxo-2H-pyridin-1-yl)acetic acid

Supplier: Matrix Scientific

041054-500MG
101850-708EA 526.15 USD
101850-708
(5-Cyclohexylcarbamoyl-2-oxo-2H-pyridin-1-yl)acetic acid
(5-Cyclohexylcarbamoyl-2-oxo-2H-pyridin-1-yl)acetic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₄
MDL Number: MFCD12028503
CAS Number: 1171917-07-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR