4-[2-(4-Fluorophenyl)acetyl]morpholine-2-carboxylic acid

Supplier: Matrix Scientific

041052-500MG
101850-704EA 363.08 USD
101850-704
4-[2-(4-Fluorophenyl)acetyl]morpholine-2-carboxylic acid
4-[2-(4-Fluorophenyl)acetyl]morpholine-2-carboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₄FNO₄
MDL Number: MFCD12028501
CAS Number: 1171917-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR