N-Azepan-4-yl-3-methoxy-N-methylbenzamide

Supplier: Matrix Scientific

041051-500MG
101850-702EA 526.15 USD
101850-702
N-Azepan-4-yl-3-methoxy-N-methylbenzamide
N-Azepan-4-yl-3-methoxy-N-methylbenzamide

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₂
MW: 262.35 g/mol
MDL Number: MFCD12028500
CAS Number: 1171917-00-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR