(1-Isobutyrylpiperidin-4-yl)acetic acid

Supplier: Matrix Scientific

041046-500MG
101850-692EA 526.15 USD
101850-692
(1-Isobutyrylpiperidin-4-yl)acetic acid
(1-Isobutyrylpiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₁H₁₉NO₃
MDL Number: MFCD12028495
CAS Number: 1171916-91-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR