(1-Pyridin-2-ylmethylpiperidin-4-yl)acetic acid

Supplier: Matrix Scientific

041045-500MG
101850-690EA 526.15 USD
101850-690
(1-Pyridin-2-ylmethylpiperidin-4-yl)acetic acid
(1-Pyridin-2-ylmethylpiperidin-4-yl)acetic acid

Hazard: Irritant

Formula: C₁₃H₁₈N₂O₂
MDL Number: MFCD12028494
CAS Number: 1171916-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR