3-(3-Aminopropyl)-3H-benzooxazol-2-one hydrochloride

Supplier: Matrix Scientific

041039-500MG
101850-678EA 526.15 USD
101850-678
3-(3-Aminopropyl)-3H-benzooxazol-2-one hydrochloride
3-(3-Aminopropyl)-3H-benzooxazol-2-one hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MDL Number: MFCD12028490
CAS Number: 1185296-13-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR