Cyclopentanecarboxylic acid(2-piperidin-4-ylethyl)amide hydrochloride
Supplier: Matrix Scientific
041037-500MG
101850-674EA
387.69
USD
101850-674
Cyclopentanecarboxylic acid(2-piperidin-4-ylethyl)amide hydrochloride
Cyclopentanecarboxylic acid(2-piperidin-4-ylethyl)amide hydrochloride
Hazard: Irritant
Formula:
C₁₃H₂₅ClN₂O |
MDL Number:
MFCD12028488 CAS Number: 1185296-32-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...