2-Methoxy-N-(2-piperidin-4-ylethyl)acetamide hydrochloride

Supplier: Matrix Scientific

041036-500MG
101850-672EA 364.62 USD
101850-672
2-Methoxy-N-(2-piperidin-4-ylethyl)acetamide hydrochloride
2-Methoxy-N-(2-piperidin-4-ylethyl)acetamide hydrochloride

Hazard: Irritant

Formula: C₁₀H₂₁ClN₂O₂
MDL Number: MFCD12028487
CAS Number: 1185302-94-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR