5-Bromo-2-furamide

Supplier: Matrix Scientific

Synonyms: 5-Bromofuran-2-carboxamide

041021-500MG
101850-630EA 450.77 USD
101850-630
5-Bromo-2-furamide
5-Bromo-2-furamide

Hazard: Irritant

Formula: C₅H₄BrNO₂
MW: 190 g/mol
MDL Number: MFCD00020926
CAS Number: 6134-61-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR