Ethyl-1-(aminocarbonyl)cyclobutanecarboxylate

Supplier: Matrix Scientific

041014-500MG
101850-614EA 290.77 USD
101850-614
Ethyl-1-(aminocarbonyl)cyclobutanecarboxylate
Ethyl-1-(aminocarbonyl)cyclobutanecarboxylate

Hazard: Irritant

Formula: C₈H₁₃NO₃
MDL Number: MFCD12028482
CAS Number: 1142198-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR