3-Hydroxy-2,2-dimethylpropanenitrile

Supplier: Matrix Scientific

041011-500MG
101850-608EA 253.85 USD
101850-608
3-Hydroxy-2,2-dimethylpropanenitrile
3-Hydroxy-2,2-dimethylpropanenitrile

Hazard: Irritant

Formula: C₅H₉NO
Boiling Pt: 86…87 °C (18 Torr)
MDL Number: MFCD12028479
CAS Number: 19295-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR