Ethyl-2-cyano-2-ethylbutanoate

Supplier: Matrix Scientific

041010-500MG
101850-606EA 193.85 USD
101850-606
Ethyl-2-cyano-2-ethylbutanoate
Ethyl-2-cyano-2-ethylbutanoate

Hazard: Irritant

Formula: C₉H₁₅NO₂
MW: 169.23 g/mol
MDL Number: MFCD00045613
CAS Number: 1619-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR