Ethyl-1-cyanocyclopentanecarboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 1-cyanocyclopentane-1-carboxylate

041008-500MG
101850-602EA 193.85 USD
101850-602
Ethyl-1-cyanocyclopentanecarboxylate
Ethyl-1-cyanocyclopentanecarboxylate

Hazard: Irritant

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD00019291
CAS Number: 28247-14-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR