1-(Methoxyacetyl)cyclopropanecarbonitrile

Supplier: Matrix Scientific

041007-500MG
101850-600EA 193.85 USD
101850-600
1-(Methoxyacetyl)cyclopropanecarbonitrile
1-(Methoxyacetyl)cyclopropanecarbonitrile

Hazard: Irritant

Formula: C₇H₉NO₂
MDL Number: MFCD12028478
CAS Number: 1142198-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR