[(3-Ethyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

041005-500MG
101850-596EA 538.46 USD
101850-596
[(3-Ethyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride
[(3-Ethyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₅H₁₀ClN₃O
MDL Number: MFCD09864216
CAS Number: 103457-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR