(Dimethylamino)(5-methyl-2-furyl)acetonitrile

Supplier: Matrix Scientific

041003-500MG
101850-592EA 193.85 USD
101850-592
(Dimethylamino)(5-methyl-2-furyl)acetonitrile
(Dimethylamino)(5-methyl-2-furyl)acetonitrile

Hazard: Irritant

Formula: C₉H₁₂N₂O
MDL Number: MFCD12028475
CAS Number: 1142198-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR