4-(Hydroxymethyl)pyrrolidin-2-one

Supplier: Matrix Scientific

041000-1G
101850-586EA 200 USD
101850-586
4-(Hydroxymethyl)pyrrolidin-2-one
4-(Hydroxymethyl)pyrrolidin-2-one

Hazard: Irritant

Formula: C₅H₉NO₂
MDL Number: MFCD11845728
CAS Number: 64320-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR