Ethyl (2-ethyl-5-oxo-1,4-diazepan-1-yl)acetate

Supplier: Matrix Scientific

040999-500MG
101850-584EA 387.69 USD
101850-584
Ethyl (2-ethyl-5-oxo-1,4-diazepan-1-yl)acetate
Ethyl (2-ethyl-5-oxo-1,4-diazepan-1-yl)acetate

Hazard: Irritant

Formula: C₁₁H₂₀N₂O₃
MDL Number: MFCD12028474
CAS Number: 1142198-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR