6-(Hydroxymethyl)pyridin-2-ol

Supplier: Matrix Scientific

040998-500MG
101850-582EA 576.92 USD
101850-582
6-(Hydroxymethyl)pyridin-2-ol
6-(Hydroxymethyl)pyridin-2-ol

Hazard: Irritant

Formula: C₆H₇NO₂
MDL Number: MFCD12028473
CAS Number: 352514-21-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR