(2,3-Dimethyl-1H-indol-5-yl)methanol

Supplier: Matrix Scientific

040997-500MG
101850-580EA 290.77 USD
101850-580
(2,3-Dimethyl-1H-indol-5-yl)methanol
(2,3-Dimethyl-1H-indol-5-yl)methanol

Hazard: Irritant

Formula: C₁₁H₁₃NO
MDL Number: MFCD12028472
CAS Number: 1071667-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR