Methyl [1-(methoxymethyl)cyclopentyl]carbamate

Supplier: Matrix Scientific

040994-500MG
101850-574EA 193.85 USD
101850-574
Methyl [1-(methoxymethyl)cyclopentyl]carbamate
Methyl [1-(methoxymethyl)cyclopentyl]carbamate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD12028470
CAS Number: 1142197-99-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR