1-(Methoxymethyl)cyclopentanecarboxamide

Supplier: Matrix Scientific

040993-500MG
101850-572EA 193.85 USD
101850-572
1-(Methoxymethyl)cyclopentanecarboxamide
1-(Methoxymethyl)cyclopentanecarboxamide

Hazard: Irritant

Formula: C₈H₁₅NO₂
MDL Number: MFCD12028469
CAS Number: 1142211-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR