9-Benzyl-6-oxa-9-azaspiro[3.6]decan-8-one

Supplier: Matrix Scientific

040989-500MG
101850-564EA 290.77 USD
101850-564
9-Benzyl-6-oxa-9-azaspiro[3.6]decan-8-one
9-Benzyl-6-oxa-9-azaspiro[3.6]decan-8-one

Hazard: Irritant

Formula: C₁₅H₁₉NO₂
MDL Number: MFCD12028465
CAS Number: 1160245-66-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR