5-(4-Aminophenyl)-2,4-dihydro-3H-pyrazol-3-one

Supplier: Matrix Scientific

040984-500MG
101850-554EA 290.77 USD
101850-554
5-(4-Aminophenyl)-2,4-dihydro-3H-pyrazol-3-one
5-(4-Aminophenyl)-2,4-dihydro-3H-pyrazol-3-one

Hazard: Irritant

Formula: C₉H₉N₃O
MDL Number: MFCD08059837
CAS Number: 103755-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR