[3-(1H-Pyrazol-1-yl)phenyl]methanol

Supplier: Matrix Scientific

Synonyms: 3-(1H-Pyrazol-1-yl)benzyl alcohol

040982-500MG
101850-550EA 187.69 USD
101850-550
[3-(1H-Pyrazol-1-yl)phenyl]methanol
[3-(1H-Pyrazol-1-yl)phenyl]methanol

Hazard: Irritant

Formula: C₁₀H₁₀N₂O
MW: 174.2 g/mol
MDL Number: MFCD06740322
CAS Number: 864068-80-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR