[1-(Methoxymethyl)cyclopentyl]amine hydrochloride

Supplier: Matrix Scientific

040976-500MG
101850-538EA 230.77 USD
101850-538
[1-(Methoxymethyl)cyclopentyl]amine hydrochloride
[1-(Methoxymethyl)cyclopentyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO
MDL Number: MFCD09864357
CAS Number: 944146-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR