[(3-Methyl-1,2,4-oxadiazol-5-yl)(phenyl)methyl]amine hydrochloride

Supplier: Matrix Scientific

040972-500MG
101850-530EA 323.08 USD
101850-530
[(3-Methyl-1,2,4-oxadiazol-5-yl)(phenyl)methyl]amine hydrochloride
[(3-Methyl-1,2,4-oxadiazol-5-yl)(phenyl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₂ClN₃O
MDL Number: MFCD12028455
CAS Number: 1184984-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR