[1-(3-Isobutyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride
Supplier: Matrix Scientific
040968-500MG
101850-522EA
300
USD
101850-522
[1-(3-Isobutyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride
[1-(3-Isobutyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride
Hazard: Irritant
Formula:
C₈H₁₆ClN₃O |
MDL Number:
MFCD09864306 CAS Number: 1039994-46-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...