[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

Supplier: Matrix Scientific

040967-500MG
101850-520EA 484.62 USD
101850-520
[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride
[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]amine hydrochloride

Hazard: Irritant

Formula: C₄H₈ClN₃O
MDL Number: MFCD12028451
CAS Number: 253196-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR