[2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

040965-500MG
101850-516EA 387.69 USD
101850-516
[2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride
[2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₄ClN₃O
MDL Number: MFCD12028449
CAS Number: 915923-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR