[2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride

Supplier: Matrix Scientific

040964-500MG
101850-514EA 484.62 USD
101850-514
[2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride
[2-(3-Pyrazin-2-yl-1,2,4-oxadiazol-5-yl)ethyl]amine hydrochloride

Hazard: Irritant

Formula: C₈H₁₀ClN₅O
MDL Number: MFCD12028448
CAS Number: 1185298-52-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR