[2-(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)ethyl]amine trifluoroacetate

Supplier: Matrix Scientific

040963-500MG
101850-512EA 338.46 USD
101850-512
[2-(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)ethyl]amine trifluoroacetate
[2-(3-Pyridin-3-yl-1,2,4-oxadiazol-5-yl)ethyl]amine trifluoroacetate

Hazard: Irritant

Formula: C₁₁H₁₁F₃N₄O₃
MDL Number: MFCD12028447
CAS Number: 1185302-86-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR