3-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

Supplier: Matrix Scientific

040956-500MG
101850-498EA 146.15 USD
101850-498
3-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride
3-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃O
MDL Number: MFCD12028440
CAS Number: 1185296-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR