2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

Supplier: Matrix Scientific

040955-500MG
101850-496EA 387.69 USD
101850-496
2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride
2-(3-Isopropyl-1,2,4-oxadiazol-5-yl)piperidine hydrochloride

Hazard: Irritant

Formula: C₁₀H₁₈ClN₃O
MDL Number: MFCD12028439
CAS Number: 1185295-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR