4-[(Methylsulfonyl)methyl]piperidine hydrochloride

Supplier: Matrix Scientific

040951-500MG
101850-488EA 387.69 USD
101850-488
4-[(Methylsulfonyl)methyl]piperidine hydrochloride
4-[(Methylsulfonyl)methyl]piperidine hydrochloride

Hazard: Irritant

Formula: C₇H₁₆ClNO₂S
MDL Number: MFCD12028435
CAS Number: 597563-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR